Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PTH Antibody (SPM604) - N-terminal, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP2451120.02MG
Description
Ensure accurate, reproducible results in Flow Cytometry, Immunohistochemistry (Paraffin), Immunofluorescence
PTH Monoclonal specifically detects PTH in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
PTH | |
Monoclonal | |
0.2mg/mL | |
Flow Cytometry 0.5 - 1 ug/million cells in 0.1 ml, Immunohistochemistry-Paraffin 0.5 - 1.0 ug/ml, Immunofluorescence 0.5 - 1.0 ug/ml | |
Parathormone, Parathyrin, parathyroid hormone, parathyroid hormone 1, PTH1 | |
Mouse | |
9 kDa | |
0.02 mg | |
Apoptosis, Cancer | |
5741 | |
Human | |
Purified |
Flow Cytometry, Immunohistochemistry (Paraffin), Immunofluorescence | |
SPM604 | |
Unconjugated | |
10mM PBS and 0.05% BSA with 0.05% Sodium Azide | |
PTH | |
A synthetic peptide from the N-terminal of human PTH polypeptide. | |
Protein A or G purified | |
RUO | |
Primary | |
Epitope of this MAb maps in the N-terminus of PTH, a hormone produced by the parathyroid gland that regulates the concentration of calcium and phosphorus in extracellular fluid. This hormone elevates blood Ca2+ levels by dissolving the salts in bone and preventing their renal excretion.It is produced in the parathyroid gland as an 84 amino acid single chain polypeptide. It can also be secreted as N-terminal truncated fragments or C-terminal fragments after intracellular degradation, as in case of hypercalcemia. Defects in this gene are a cause of familial isolated hypoparathyroidism (FIH); also called autosomal dominant hypoparathyroidism or autosomal dominant hypocalcemia. FIH is characterized by hypocalcemia and hyperphosphatemia due to inadequate secretion of parathyroid hormone. Symptoms are seizures, tetany and cramps. FIH exist both as autosomal dominant and recessive forms of hypoparathyroidism. | |
Store at 4C. | |
IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction