Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PTH Monoclonal Antibody (OTI1C6), TrueMAB™, OriGene
SDP

Catalog No. 501678627
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-167-8627 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-167-8627 Supplier OriGene Supplier No. TA800944
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Parathyroid hormone (PTH) is secreted by parathyroid cells. PTH increases blood Ca2+ levels by dissolving the salts in bone and preventing their renal excretion. Most PTH is secreted in its intact, mature 9.5 kDa form, amino acids 1-84; however, it can also be secreted as N-terminal truncated fragments or C-terminal fragments after intracellular degradation, as in the case of hypercalcemia.

Specifications

Antigen PTH
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI1C6
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene PTH
Gene Accession No. P01270
Gene Alias PTH1
Gene Symbols PTH
Host Species Mouse
Immunogen Human recombit protein fragment corresponding to amino acids 32-115 of human PTH produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5741
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.