Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PTH Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA583914
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA583914 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA583914 Supplier Invitrogen™ Supplier No. PA583914
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KSVKKRSVSE IQLMHNLGKH LNSMERVEWL RKKLQDVHNF VALGAPLAPR DAGSQRPRKK EDNVLVESHE KSLGEADKAD VNVLTKAK Highest antigen sequence indentity to the following orthologs: Rat - 73%, Mouse - 69%.

This gene encodes a member of the parathyroid family of proteins. The encoded preproprotein is proteolytically processed to generate a protein that binds to the parathyroid hormone/parathyroid hormone-related peptide receptor and regulates blood calcium and phosphate levels. Excess production of the encoded protein, known as hyperparathyroidism, can result in hypercalcemia and kidney stones. On the other hand, defective processing of the encoded protein may lead to hypoparathyroidism, which can result in hypocalcemia and numbness.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PTH
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PTH
Gene Accession No. P01270
Gene Alias hypothalamic parathyroid hormone; parathormone; Parathyrin; parathyroid hormone; parathyroid hormone 1; preproparathyroid hormone; prepro-PTH; PTH; PTH-(1-84); Pth1; Pthp; Pthr1
Gene Symbols PTH
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PTH. Recombinant protein control fragment (Product #RP-106194).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5741
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.