Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PTH Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5110238
Description
Immunogen sequence: LTKSDGKSVK KRSVSEIQLM HNLGKHLNSM ERVEWLRKKL QDVHNFVALG APLAPRDAGS QRPRKKEDNV LVESHEKSLG EADKADVNVL TKAKSQ.
This gene encodes a member of the parathyroid family of proteins. The encoded preproprotein is proteolytically processed to generate a protein that binds to the parathyroid hormone/parathyroid hormone-related peptide receptor and regulates blood calcium and phosphate levels. Excess production of the encoded protein, known as hyperparathyroidism, can result in hypercalcemia and kidney stones. On the other hand, defective processing of the encoded protein may lead to hypoparathyroidism, which can result in hypocalcemia and numbness.
Specifications
PTH | |
Polyclonal | |
Unconjugated | |
PTH | |
hypothalamic parathyroid hormone; parathormone; Parathyrin; parathyroid hormone; parathyroid hormone 1; preproparathyroid hormone; prepro-PTH; PTH; PTH-(1-84); Pth1; Pthp; Pthr1 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
5741 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
0.99 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P01270 | |
PTH | |
Recombinant fusion protein containing a sequence corresponding to amino acids 20-115 of human Parathyroid Hormone (Parathyroid Hormone (PTH)) (NP_0003061). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction