Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PTH Rabbit anti-Human, Mouse, Rat, Clone: 5X6R6, Novus Biologicals™

Rabbit Monoclonal Antibody
$205.50 - $458.50
Specifications
Antigen | PTH |
---|---|
Clone | 5X6R6 |
Dilution | Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:50 - 1:200 |
Applications | Western Blot, Immunohistochemistry, Immunofluorescence |
Classification | Monoclonal |
Description
PTH Monoclonal antibody specifically detects PTH in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, ImmunofluorescenceSpecifications
PTH | |
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:50 - 1:200 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
Parathormone, Parathyrin, parathyroid hormone, parathyroid hormone 1, PTH1 | |
A synthetic peptide corresponding to a sequence within amino acids 1-100 of human PTH (PTH) (P01270). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
5X6R6 | |
Western Blot, Immunohistochemistry, Immunofluorescence | |
Unconjugated | |
Rabbit | |
Apoptosis, Biologically Active Proteins, Cancer | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
5741 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title