Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PTH1R Recombinant Rabbit Monoclonal Antibody (1G7)
GREENER_CHOICE

Catalog No. PIMA538295
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA538295 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA538295 Supplier Invitrogen™ Supplier No. MA538295
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

This antibody has been tested in direct-ELISA.

Parathyroid hormone receptor 1 is a member of the G-protein coupled receptor family. This protein is a receptor for parathyroid hormone (PTH) and for parathyroid hormone-like hormone (PTHLH). The activity of this receptor is mediated by G proteins which activate adenylyl cyclase and also a phosphatidylinositol-calcium second messenger system. Defects in this receptor are known to be the cause of Jansen's metaphyseal chondrodysplasia (JMC), chondrodysplasia Blomstrand type (BOCD), as well as enchodromatosis. Parathyroid hormone receptor 1 has been reported to be expressed in bone, breast, brain, colon, GI, kidney, liver, placenta, skin, umbilical cord, and uterus.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PTH1R
Applications Western Blot
Classification Recombinant Monoclonal
Clone 1G7
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol, 0.05% BSA and 0.02% sodium azide; pH 7.4
Gene Pth1r
Gene Accession No. Q03431
Gene Alias MGC138426; MGC138452; Parathormone; Parathyrin; parathyroid hormone 1 receptor; parathyroid hormone receptor; parathyroid hormone receptor 1; parathyroid hormone/parathyroid hormone-related peptide receptor; parathyroid hormone/parathyroid hormone-related protein receptor; PFE; PPR; PTH; PTH/PTHr receptor; PTH/PTHrP receptor; PTH/PTHrP type I receptor; PTH1 receptor; PTH1R; Pthr; Pthr1; PTHrel; PTH-related peptide receptor; PTHrP-R; PTHrP-receptor; seven transmembrane helix receptor
Gene Symbols Pth1r
Host Species Rabbit
Immunogen A synthesized peptide derived from human Parathyroid Hormone Receptor 1.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5745
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.