Learn More
PTH1R Rabbit anti-Human, Polyclonal, ProteinTech

Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 291151AP150UL

Description
Aliquoting is unnecessary for -20°C storage.
PTH1R Polyclonal antibody specifically detects PTH1R in Human samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
PTH1R | |
Polyclonal | |
Unconjugated | |
Pth1r | |
MGC138426; MGC138452; Parathormone; Parathyrin; parathyroid hormone 1 receptor; parathyroid hormone receptor; parathyroid hormone receptor 1; parathyroid hormone/parathyroid hormone-related peptide receptor; parathyroid hormone/parathyroid hormone-related protein receptor; PFE; PPR; PTH; PTH/PTHr receptor; PTH/PTHrP receptor; PTH/PTHrP type I receptor; PTH1 receptor; PTH1R; Pthr; Pthr1; PTHrel; PTH-related peptide receptor; PTHrP-R; PTHrP-receptor; seven transmembrane helix receptor | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
5745 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
700 μg/mL | |
PBS with 50% glycerol and 0.02% sodium azide | |
Q03431 | |
Pth1r | |
PTH1R Fusion Protein Ag30762 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.