Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PTH2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA569943
Description
This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 93%; Rat: 100%.
PTH2 gene ontology annotations related to this gene include neuropeptide signaling pathway.
Specifications
PTH2 | |
Polyclonal | |
Unconjugated | |
PTH2 | |
parathyroid hormone 2; PTH2; RGD1559447; Tifp39; Tip39; TIPF39; tuberoinfundibular 39 residue protein; tuberoinfundibular peptide of 39 residues; tuberoinfundibular peptide of 39 residues (TIP39) preprohormone | |
Rabbit | |
Affinity Chromatography | |
RUO | |
113091 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q96A98 | |
PTH2 | |
synthetic peptide directed towards the middle region of human PTH2. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction