Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PTH2R Monoclonal Antibody (C1), Invitrogen™
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA543618
This item is not returnable.
View return policy
Description
PTHR2 is a specific receptor for parathyroid hormone. The activity of this receptor is mediated by g proteins which activate adenylyl cyclase. PTHR2 may be responsible for PTH effects in a number of physiological systems, and may also play a significant role in pancreatic function. PTHR2 presence in neurons indicates that it may function as a neurotransmitter receptor.
Specifications
PTH2R | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.05% ProClin 300 | |
P49190 | |
Pth2r | |
Recombinant PTHR2 (Asp27-Tyr145) expressed in E. Coli. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG2b κ |
Immunohistochemistry (Paraffin), Western Blot | |
C1 | |
Unconjugated | |
Pth2r | |
Hpth2 receptor; parathyroid hormone 2 receptor; parathyroid hormone receptor; parathyroid hormone receptor 2; PTH2 receptor; Pth2r; PTHR2 | |
Mouse | |
Protein A/G | |
RUO | |
5746 | |
-20°C | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction