Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PTH2R Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA570662
Description
This target displays homology in the following species: Cow: 79%; Guinea Pig: 86%; Human: 100%.
PTHR2 is a specific receptor for parathyroid hormone. The activity of this receptor is mediated by g proteins which activate adenylyl cyclase. PTHR2 may be responsible for PTH effects in a number of physiological systems, and may also play a significant role in pancreatic function. PTHR2 presence in neurons indicates that it may function as a neurotransmitter receptor.
Specifications
PTH2R | |
Polyclonal | |
Unconjugated | |
Pth2r | |
Hpth2 receptor; parathyroid hormone 2 receptor; parathyroid hormone receptor; parathyroid hormone receptor 2; PTH2 receptor; Pth2r; PTHR2 | |
Rabbit | |
Affinity chromatography | |
RUO | |
5746 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
P49190 | |
Pth2r | |
synthetic peptide directed towards the C-terminal region of human PTH2R. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction