Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PTHLH Monoclonal Antibody (3979)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA536294
Description
PTHLH Monoclonal Antibody for Western Blot, IHC (P), Flow, ELISA
The protein encoded by this gene is a member of the parathyroid hormone family. This hormone regulates endochondral bone development and epithelial-mesenchymal interactions during the formation of the mammary glands and teeth. This hormone is involved in lactation possibly by regulating the mobilization and transfer of calcium to the milk. The receptor of this hormone, PTHR1, is responsible for most cases of humoral hypercalcemia of malignancy. Four alternatively spliced transcript variants encoding two distinct isoforms have been observed. There is also evidence for alternative translation initiation from non-AUG (CUG and GUG) start sites, in-frame and downstream of the initiator AUG codon, to give rise to nuclear forms of this hormone.
Specifications
PTHLH | |
Monoclonal | |
1 mg/mL | |
PBS with 0.1% sodium azide | |
P12272 | |
PTHLH | |
23 amino acid peptide (corresponding to amino acids 146-169) of the hPTHPTHrP receptor. | |
100 μL | |
Primary | |
Human | |
Antibody | |
Ig |
ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
3979 | |
Unconjugated | |
PTHLH | |
BDE2; calciotropic hormone, growth factor; HHM; osteostatin; parathyroid hormone like hormone; parathyroid hormone related peptide; parathyroid hormone-like hormone; parathyroid hormone-like hormone preproprotein; parathyroid hormone-like peptide; parathyroid hormone-like protein; parathyroid hormone-like related protein; parathyroid hormone-related peptide; parathyroid hormone-related protein; parathyroid-like peptide; PLP; PTHLH; PTH-like; PTHR; PTH-related peptide; PTH-related protein; Pthrp; PTH-rP; PTHrP[107-139]; PTHrP[1-36]; PTHrP[38-94] | |
Mouse | |
Protein G | |
RUO | |
5744 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction