Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PTIP Antibody, HRP, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB100577H
Description
PTIP Polyclonal antibody specifically detects PTIP in Human samples. It is validated for Western BlotSpecifications
PTIP | |
Polyclonal | |
Western Blot | |
CAGF28, PACIP1, PAX interacting (with transcription-activation domain) protein 1, PAX transactivation activation domain-interacting protein, PAX transcription activation domain interacting protein 1 like, PAX-interacting protein 1, PAXIP1LCAGF29, protein encoded by CAG trinucleotide repeats, PTIPFLJ41049, TNRC2 | |
The immunogen recognized by this antibody maps to a region between residues 1 and 50 of human PAX transcription activation domain interacting protein 1 using the numbering given in Jowsey et akl., 2004, J. Biol. Chem. 279(53):55562-55569 (GeneID 22976). | |
0.1 mL | |
Cancer, DNA Repair | |
22976 | |
Store at 4C in the dark. | |
IgG |
Western Blot | |
HRP | |
PBS | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction