Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PTIP Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $646.00
Specifications
Antigen | PTIP |
---|---|
Dilution | Western Blot 0.4 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin 1:10-1:20 |
Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
PTIP Polyclonal specifically detects PTIP in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
PTIP | |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
Human | |
CAGF28, PACIP1, PAX interacting (with transcription-activation domain) protein 1, PAX transactivation activation domain-interacting protein, PAX transcription activation domain interacting protein 1 like, PAX-interacting protein 1, PAXIP1LCAGF29, protein encoded by CAG trinucleotide repeats, PTIPFLJ41049, TNRC2 | |
PAXIP1 | |
IgG | |
Affinity Purified | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Western Blot 0.4 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin 1:10-1:20 | |
Polyclonal | |
Rabbit | |
Cancer, DNA Repair | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
22976 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:EEEEEVENEEQDSQNEGSTDEKSSPASSQEGSPSGDQQFSPKSNTEKSKGELMFDDSSDSSPEKQERNLNWTPAEVPQLAAAKRRLPQGKEPGLINLCANVPPVPGNILPPEVRGNLMAAGQNLQSSERS | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title