Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PTIP Polyclonal Antibody, Bethyl Laboratories

Catalog No. 501572642
Encompass_Preferred
Change view
Click to view available options
Quantity:
20 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-157-2642 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-157-2642 Supplier Bethyl Laboratories, Inc Supplier No. A300370AT
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt.

This gene is a member of the paired box gene family and encodes a nuclear protein with six BRCT domains. This protein plays a critical role in maintaining genome stability, condensation of chromatin and progression through mitosis.

Specifications

Antigen PTIP
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation TBS with 0.1% BSA and 0.09% sodium azide
Gene PAXIP1
Gene Accession No. Q6ZW49
Gene Alias CAGF28, CAGF29, FLJ41049, PACIP1, PAXIP1L, PTIP, TNRC2, PAX transcription activation domain interacting protein 1 like; PAX-interacting protein 1; protein encoded by CAG trinucleotide repeats
Gene Symbols PAXIP1
Host Species Rabbit
Immunogen Between 1 and 50
Purification Method Antigen affinity chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22976
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.