Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PTK5 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501987227
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-7227 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-7227 Supplier Bioss Supplier No. BS6382R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

FRK (Fyn-related kinase), also known as Rak. It is an epithelial tissue-specific kinase. The human FRK gene maps to chromosome 6q21-q22.3 and encodes a 505 amino acid protein. The protein belongs to the TYR family of protein kinases. This tyrosine kinase is a nuclear protein and may function during G1 and S phase of the cell cycle and suppress growth.

Specifications

Antigen PTK5
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene FRK
Gene Accession No. P42685
Gene Alias B-cell src-homology tyrosine kinase; Beta-cell Src-homology tyrosine kinase; Bsk; BSK/IYK; C85044; EC 2.7.1.112; EC 2.7.10.2; Frk; fyn related Src family tyrosine kinase; FYN-related kinase; fyn-related Src family tyrosine kinase; Gask; Gastrointestinal-associated kinase; Gastrointestinal-associated tyrosine kinase; GTK; Intestine tyrosine kinase; Iyk; Nuclear tyrosine protein kinase RAK; Protein-tyrosine kinase 5; PTK5; PTK5 protein tyrosine kinase 5; RAK; src related tyrosine kinase; Tyrosine-protein kinase FRK
Gene Symbols FRK
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human FRK/PTK5.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2444
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.