Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PTK7/CCK4 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$646.00
Specifications
Antigen | PTK7/CCK4 |
---|---|
Dilution | Western Blot 0.04 - 0.4 ug/ml, Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500 |
Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
PTK7/CCK4 Polyclonal specifically detects PTK7/CCK4 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
PTK7/CCK4 | |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
Human | |
Q13308 | |
5754 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:TAIVFIKQPSSQDALQGRRALLRCEVEAPGPVHVYWLLDGAPVQDTERRFAQGSSLSFAAVDRLQDSGTFQCVARDDVTGEEARSANASFNIKWIEAGPVVLKHPASEAEIQPQTQVTLRCHIDGHPRPTYQWFR | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Western Blot 0.04 - 0.4 ug/ml, Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
Polyclonal | |
Rabbit | |
Protein Kinase | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
CCK-4, CCK4Colon carcinoma kinase 4, colon carcinoma kinase-4, Protein-tyrosine kinase 7, Pseudo tyrosine kinase receptor 7, PTK7 protein tyrosine kinase 7, Tyrosine-protein kinase-like 7 | |
PTK7 | |
IgG | |
Affinity Purified | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title