Learn More
PTK7/CCK4 Antibody (OTI2E7), Alexa Fluor™ 647, Novus Biologicals™
SDP
Mouse Monoclonal Antibody
Supplier:  Novus Biologicals NBP273708AF647
Catalog No. N273708A647
Only null left
Add to Cart
Description
Description
PTK7/CCK4 Monoclonal specifically detects PTK7/CCK4 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence.
Specifications
Specifications
PTK7/CCK4
Monoclonal
Alexa Fluor 647
50mM Sodium Borate with 0.05% Sodium Azide
PTK7
Full length recombinant protein of human PTK7/CCK4 (NP_690619) produced in HEK293T cells.
0.1 mL
Primary
Human, Mouse
IgG1
Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence
OTI2E7
Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence
CCK-4, CCK4Colon carcinoma kinase 4, colon carcinoma kinase-4, Protein-tyrosine kinase 7, Pseudo tyrosine kinase receptor 7, PTK7 protein tyrosine kinase 7, Tyrosine-protein kinase-like 7
Mouse
Affinity Purified
Protein Kinase
5754.0
Store at 4°C in the dark.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.