Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PTK7/CCK4 Antibody (OTI2E7), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP201486
Description
PTK7/CCK4 Monoclonal specifically detects PTK7/CCK4 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence.Specifications
| PTK7/CCK4 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:200-500, Flow Cytometry 1:100, Immunocytochemistry/Immunofluorescence 1:100 | |
| PTK7 | |
| Full length human recombinant protein of human PTK7(NP_690619) produced in HEK293T cell. | |
| 0.1 mL | |
| Protein Kinase | |
| 5754 | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
| OTI2E7 | |
| Unconjugated | |
| CCK-4, CCK4Colon carcinoma kinase 4, colon carcinoma kinase-4, Protein-tyrosine kinase 7, Pseudo tyrosine kinase receptor 7, PTK7 protein tyrosine kinase 7, Tyrosine-protein kinase-like 7 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction