Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PTK7/CCK4 Antibody (OTI2E7), PE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273708PE
Description
PTK7/CCK4 Monoclonal specifically detects PTK7/CCK4 in Human, Mouse samples. It is validated for Flow Cytometry.Specifications
PTK7/CCK4 | |
Monoclonal | |
PE | |
PBS with 0.05% Sodium Azide | |
PTK7 | |
Full length recombinant protein of human PTK7/CCK4 (NP_690619) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human, Mouse | |
IgG1 |
Flow Cytometry | |
OTI2E7 | |
Flow Cytometry | |
CCK-4, CCK4Colon carcinoma kinase 4, colon carcinoma kinase-4, Protein-tyrosine kinase 7, Pseudo tyrosine kinase receptor 7, PTK7 protein tyrosine kinase 7, Tyrosine-protein kinase-like 7 | |
Mouse | |
Affinity Purified | |
Protein Kinase | |
5754.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction