Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PTK7 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA520425
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA520425 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA520425 Supplier Invitrogen™ Supplier No. PA520425
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

A suggested positive control is human colon tissue lysate. PA5-20425 can be used with blocking peptide PEP-0542.

CCK4 may function as a cell adhesion molecule. Although it belongs to the insulin receptor subfamily of the Tyr protein kinases, it likely lacks the catalytic activity of a tyrosine kinase. It may be connected to the pathophysiology of colon carcinomas and/or may represent a tumor progression marker. This Type I membrane protein is highly expressed in lung, liver, pancreas, kidney, placenta and melanocytes, but weakly expressed in thyroid gland, ovary, brain, heart and skeletal muscle, and not in colon. It is also expressed in erythroleukemia cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PTK7
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene Ptk7
Gene Accession No. Q13308, Q8BKG3
Gene Alias 8430404F20Rik; CCK4; CCK-4; chz; Colon carcinoma kinase 4; inactive tyrosine-protein kinase 7; mPTK7/CCK4; protein chuzhoi; protein tyrosine kinase 7; protein tyrosine kinase 7 (inactive); protein-tyrosine kinase 7; Pseudo tyrosine kinase receptor 7; Ptk7; PTK7 protein tyrosine kinase 7; receptor tyrosine kinase-like protein; serum response factor; Srf; Tyrosine-protein kinase-like 7
Gene Symbols Ptk7
Host Species Rabbit
Immunogen A 15 amino acid peptide from near the amino terminus of human PTK7.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 301242, 5754, 71461
Target Species Human, Mouse, Rat
Content And Storage Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.