Learn More
Invitrogen™ PTK9 Monoclonal Antibody (7C11)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA517194
Description
MA5-17194 targets TWF1 in indirect ELISA, FACS, ICC, IHC, IF and WB applications and shows reactivity with Human samples. The MA5-17194 immunogen is purified recombinant fragment of human TWF1 (amino acids: 335-384 ) expressed in E. Coli. MA5-17194 detects TWF1 which has a predicted molecular weight of approximately 40.3kDa.
This gene encodes twinfilin, an actin monomer-binding protein conserved from yeast to mammals. Studies of the mouse counterpart suggest that this protein may be an actin monomer-binding protein, and its localization to cortical G-actin-rich structures may be regulated by the small GTPase RAC1.
Specifications
PTK9 | |
Monoclonal | |
1 mg/mL | |
PBS with 0.05% sodium azide | |
Q12792 | |
TWF1 | |
Purified recombinant fragment of human TWF1 (amino acids: 335-384 ) expressed in E. Coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
7C11 | |
Unconjugated | |
TWF1 | |
A6; A6 protein tyrosine kinase; actin monomer-binding protein; protein A6; Protein tyrosine kinase 9; PTK9; PTK9 protein tyrosine kinase 9; TWF1; twinfilin; twinfilin actin binding protein 1; twinfilin actin-binding protein 1; twinfilin, actin-binding protein, homolog 1; twinfilin, actin-binding protein, homolog 1 (Drosophila); twinfilin-1 | |
Mouse | |
Protein G | |
RUO | |
5756 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.