Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PTK9 Antibody (OTI5C12), Biotin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273710B
Description
PTK9 Monoclonal specifically detects PTK9 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Knockout Validated.Specifications
PTK9 | |
Monoclonal | |
Biotin | |
PBS with 0.05% Sodium Azide | |
TWF1 | |
Recombinant protein fragment corresponding to amino acids 140-384 of human PTK9 (NP_002813) produced in E.coli. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG2b |
Western Blot, Immunohistochemistry, Immunoassay | |
OTI5C12 | |
Western Blot, Immunohistochemistry, Knockout Validated | |
A6, A6 protein tyrosine kinase, Protein A6, Protein tyrosine kinase 9MGC23788, PTK9 protein tyrosine kinase 9, PTK9MGC41876, twinfilin, actin-binding protein, homolog 1 (Drosophila), twinfilin-1 | |
Mouse | |
Affinity Purified | |
Phospho Specific, Protein Kinase | |
5756.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction