Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PTK9 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA530949
Description
Recommended positive controls: A549, HeLa, HepG2, HCT116. Predicted reactivity: Mouse (92%), Rat (94%), Xenopus laevis (80%), Chicken (90%), Bovine (95%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene encodes twinfilin, an actin monomer-binding protein conserved from yeast to mammals. Studies of the mouse counterpart suggest that this protein may be an actin monomer-binding protein, and its localization to cortical G-actin-rich structures may be regulated by the small GTPase RAC1.
Specifications
| PTK9 | |
| Polyclonal | |
| Unconjugated | |
| TWF1 | |
| A6; A6 protein tyrosine kinase; actin monomer-binding protein; protein A6; Protein tyrosine kinase 9; PTK9; PTK9 protein tyrosine kinase 9; TWF1; twinfilin; twinfilin actin binding protein 1; twinfilin actin-binding protein 1; twinfilin, actin-binding protein, homolog 1; twinfilin, actin-binding protein, homolog 1 (Drosophila); twinfilin-1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 5756 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
| Q12792 | |
| TWF1 | |
| Recombinant protein encompassing a sequence within the center region of human PTK9. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction