Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PTK9 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA553729
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA553729 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA553729 Supplier Invitrogen™ Supplier No. PA553729
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: AEEELRQIKI NEVQTDVGVD TKHQTLQGVA FPISREAFQA LEKLNNRQLN YVQLEIDIKN EIIILANTTN TELKDLPKRI PKDSARYHFF LYKHSHEGDY Highest antigen sequence identity to the following orthologs: Mouse - 92%, Rat - 93%.

This gene encodes twinfilin, an actin monomer-binding protein conserved from yeast to mammals. Studies of the mouse counterpart suggest that this protein may be an actin monomer-binding protein, and its localization to cortical G-actin-rich structures may be regulated by the small GTPase RAC1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PTK9
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TWF1
Gene Accession No. Q12792, Q5RJR2, Q91YR1
Gene Alias A6; A6 protein tyrosine kinase; actin monomer-binding protein; protein A6; Protein tyrosine kinase 9; PTK9; PTK9 protein tyrosine kinase 9; TWF1; twinfilin; twinfilin actin binding protein 1; twinfilin actin-binding protein 1; twinfilin, actin-binding protein, homolog 1; twinfilin, actin-binding protein, homolog 1 (Drosophila); twinfilin-1
Gene Symbols TWF1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PTK9. Recombinant protein control fragment (Product #RP-91452).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 19230, 315265, 5756
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.