Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PTMA Polyclonal Antibody

Goat Polyclonal Antibody
Supplier: Invitrogen™ PA5142436
This item is not returnable.
View return policy
Description
Additional Information: This antibody is expected to recognize both reported isoforms (NP_001092755.1; NP_002814.3). This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:1,000.
The thymus gland produces several hormones or hormone-like substances which are derived from a polypeptide precursor containing (in the rat) 113 amino acids and known as prothymosin-alpha. A peptide containing 28 amino acid residues, named thymosin-alpha-1, was originally isolated from calf thymosin fraction 5 and shown to restore various aspects of immune function in several in vitro and invivo test systems. Thymosin-alpha-1 was subsequently isolated from a similar fraction from human thymosin and reported to have the same amino acid sequence as bovine thymosin-alpha-1. In 2003, Jiang et al. identified a pathway that regulates mitochondria-initiated caspase activity. In this pathway, putative human HLA class II-associated protein (PHAP) promoted caspase-9 activation after apoptosome formation, whereas prothymosin-alpha-1 negatively regulated caspase-9 activation by inhibiting apoptosome formation.
Specifications
PTMA | |
Polyclonal | |
Unconjugated | |
PTMA | |
alpha-prothymosin; gene sequence 28; prothymosin alpha; prothymosin alpha protein; Prothymosin alpha, N-terminally processed; prothymosin, alpha; prothymosin-alpha; PTMA; Thym; thymosin; Thymosin alpha; Thymosin alpha-1; TMSA; TMSB4; TMSB4X | |
Goat | |
Ammonium sulfate precipitation | |
RUO | |
5757 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
P06454 | |
PTMA | |
Peptide with sequence C-DVDTKKQKTDEDD. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction