Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PTMS Rabbit anti-Human, Polyclonal, Proteintech
SDP

Catalog No. 501727718 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-172-7718 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-172-7718 Supplier Proteintech Group Inc Supplier No. 121061AP150UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Product No. 12106-1-AP is a rabbit polyclonal antibody raised against the full-length of human PTMS. The MW of this protein is 12 kDa, and this antibody specifically recognizes the 12 kDa protein.

PTMS may mediate immune function by blocking the effect of prothymosin alpha which confers resistance to certain opportunistic infections.

Specifications

Antigen PTMS
Applications Immunocytochemistry, Immunofluorescence, Western Blot
Classification Polyclonal
Concentration 0.13 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene PTMS
Gene Accession No. P20962
Gene Alias ParaT, parathymosin, PTMS
Gene Symbols PTMS
Host Species Rabbit
Immunogen PTMS Fusion Protein Ag2751
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5763
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.