Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PTOV1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA578503
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA578503 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA578503 Supplier Invitrogen™ Supplier No. PA578503
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Predicted Reactivity: Mouse (94%), Rat (94%), Bovine (96%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

PTOV1 may activate transcription. Required for nuclear translocation of FLOT1. Promotes cell proliferation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PTOV1
Applications Immunocytochemistry
Classification Polyclonal
Concentration 2.57 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.01% thimerosal; pH 7
Gene PTOV1
Gene Accession No. Q86YD1
Gene Alias 1110030G05Rik; ACID2; Activator interaction domain-containing protein 2; AU041779; Gcap3; granule cell antiserum positive 3; PP642; prostate tumor over expressed gene 1; prostate tumor overexpressed 1; prostate tumor-overexpressed gene 1 protein; Prostate tumor-overexpressed gene 1 protein homolog; Ptov1; PTOV-1; UNQ6127/PRO20092
Gene Symbols PTOV1
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human PTOV1. The exact sequence is proprietary.
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 53635
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.