Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PTOV1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA594928
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA594928 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA594928 Supplier Invitrogen™ Supplier No. PA594928
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: Rat Brain Tissue, Rat Kidney Tissue. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.

PTOV1 may activate transcription. Required for nuclear translocation of FLOT1. Promotes cell proliferation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PTOV1
Applications Western Blot
Classification Polyclonal
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation Antibody with 5mg BSA, 0.2mg Na2PO4, 0.9mg NaCl and 0.05mg sodium azide
Gene PTOV1
Gene Accession No. Q5U2W6, Q86YD1
Gene Alias 1110030G05Rik; ACID2; Activator interaction domain-containing protein 2; AU041779; Gcap3; granule cell antiserum positive 3; PP642; prostate tumor over expressed gene 1; prostate tumor overexpressed 1; prostate tumor-overexpressed gene 1 protein; Prostate tumor-overexpressed gene 1 protein homolog; Ptov1; PTOV-1; UNQ6127/PRO20092
Gene Symbols PTOV1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence at the C-terminus of human PTOV1 (385-414aa NFVNGIRRVIANQQQVLQRNLEQEQQQRGM).
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 292888, 53635
Target Species Human, Rat
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.