Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PTOV1 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501980735
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-0735 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-0735 Supplier Bioss Supplier No. BS10171R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PTOV1 may activate transcription. Required for nuclear translocation of FLOT1. Promotes cell proliferation.

Specifications

Antigen PTOV1
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene PTOV1
Gene Accession No. Q86YD1
Gene Alias 1110030G05Rik; ACID2; Activator interaction domain-containing protein 2; AU041779; Gcap3; granule cell antiserum positive 3; PP642; prostate tumor over expressed gene 1; prostate tumor overexpressed 1; Prostate tumor-overexpressed gene 1 protein; Prostate tumor-overexpressed gene 1 protein homolog; Ptov1; PTOV-1; UNQ6127/PRO20092
Gene Symbols PTOV1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human PTOV1.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 53635
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.