Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PTP1B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5109252
Change view
Click to view available options
Quantity:
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5109252 100 μL
PIPA5109234 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PIPA5109252 Supplier Invitrogen™ Supplier No. PA5109252
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Control: Mouse prostate tissue, SiHa, HUVEC, LOVO, human tonsil tissue, human colon cancer tissue, human kidney tissue, human placenta tissue.

PTP1B (PTPN1) is a tyrosine-protein phosphatase which acts as a regulator of endoplasmic reticulum unfolded protein response. It mediates dephosphorylation of EIF2AK3/PERK and in turn inactivates the protein kinase activity of EIF2AK3/PERK. PTP1B also plays a role in CKII- and p60c-src-induced signal transduction cascades. It regulates the EFNA5-EPHA3 signaling pathway which modulates cell reorganization and cell-cell repulsion. PTP1B also regulates the hepatocyte growth factor receptor signaling pathway through dephosphorylation of MET.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PTP1B
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4
Gene PTPN1
Gene Accession No. P18031
Gene Alias CPTP1; EC 3.1.3.48; EGK_02300; hgnc9642; KIAA0283; protein tyrosine phosphatase 1B; protein tyrosine phosphatase non-receptor type 1; protein tyrosine phosphatase, non-receptor type 1; protein tyrosine phosphatase, placental; protein-protein-tyrosine phosphatase HA2; protein-tyrosine phosphatase 1B; Protein-tyrosine phosphatase HA2; PTN1; Ptp; PTP1B; PTP-1B; ptp1b protein; PTP-HA2; Ptpn1; RP5-1121H13.2; RPTPrho; tyrosine phosphatase 1b; tyrosine-phosphatase; Tyrosine-protein phosphatase non-receptor type 1; wu:fk54h03
Gene Symbols PTPN1
Host Species Rabbit
Immunogen Synthetic peptide within Human PTP1B aa 1-50.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5770
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles, store in dark
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.