Learn More
PTP1B Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A302297A

Description
The recommended shelf life for this product is 1 year from date of receipt.
Ferritin is a ubiquitous and highly conserved protein which plays a major role in iron homeostasis. It is a holoenzyme shell (∽450 kDa) consisting of 24 subunits of two types, H (heavy) and L (light), and capable of storing up to 4,500 atoms of ferric iron. Depending on the tissue type and physiologic status of the cell, the ratio of H to L subunits in ferritin can vary widely. It can be viewed not only as part of a group of iron regulatory proteins that include transferrin and the transferrin receptor, but also as a member of the protein family that orchestrates the cellular defense against stress and inflammation. Ferritin is found in the liver, spleen, kidney and heart. Only a small amount is found in the blood. The blood level of ferritin serves as an indicator of the amount of iron stored in the body.Specifications
PTP1B | |
Polyclonal | |
Unconjugated | |
PTPN1 | |
PTP1B, hgnc9642, PTN1, ptpn1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
5770 | |
4° C | |
Liquid |
Immunoprecipitation | |
1 mg/ml | |
phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
P18031 | |
PTPN1 | |
Between 275 and 325 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.