Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PTP1B Rabbit anti-Human, Polyclonal, Bethyl Laboratories

Catalog No. 501562250
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-156-2250 10 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-156-2250 Supplier Bethyl Laboratories, Inc Supplier No. A302298AT
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt.

Ferritin is a ubiquitous and highly conserved protein which plays a major role in iron homeostasis. It is a holoenzyme shell (∽450 kDa) consisting of 24 subunits of two types, H (heavy) and L (light), and capable of storing up to 4,500 atoms of ferric iron. Depending on the tissue type and physiologic status of the cell, the ratio of H to L subunits in ferritin can vary widely. It can be viewed not only as part of a group of iron regulatory proteins that include transferrin and the transferrin receptor, but also as a member of the protein family that orchestrates the cellular defense against stress and inflammation. Ferritin is found in the liver, spleen, kidney and heart. Only a small amount is found in the blood. The blood level of ferritin serves as an indicator of the amount of iron stored in the body.

Specifications

Antigen PTP1B
Applications Immunoprecipitation
Classification Polyclonal
Concentration 1 mg/ml
Conjugate Unconjugated
Formulation phosphate, tris citrate with 0.09% sodium azide; pH 7-8
Gene PTPN1
Gene Accession No. P18031
Gene Alias PTP1B, hgnc9642, PTN1, ptpn1
Gene Symbols PTPN1
Host Species Rabbit
Immunogen Between 385 and 435
Purification Method Antigen affinity chromatography
Quantity 10 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5770
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.