Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PTP1B/PTPN1 Antibody (001), Alexa Fluor™ 700, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289399AF700
Description
PTP1B/PTPN1 Monoclonal specifically detects PTP1B/PTPN1 in Human samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence.Specifications
PTP1B/PTPN1 | |
Monoclonal | |
Alexa Fluor 700 | |
EC 3.1.3.48, protein tyrosine phosphatase 1B, protein tyrosine phosphatase, non-receptor type 1, Protein-tyrosine phosphatase 1B, PTP-1B, PTP1Bprotein tyrosine phosphatase, placental, tyrosine-protein phosphatase non-receptor type 1 | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human PTP1B/PTPN1 (NP_002818.1; Glu 2-Asn 321). | |
0.1 mL | |
Phospho Specific | |
5770 | |
Store at 4°C in the dark. | |
IgG |
ELISA, Immunofluorescence | |
1 | |
50 mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction