Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PTP1B/PTPN1 Antibody (226), Janelia Fluor™ 549, Novus Biologicals™
SDP

Catalog No. NB307734 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB307734 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB307734 Supplier Novus Biologicals Supplier No. NBP289400JF549
Only null left
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

PTP1B/PTPN1 Monoclonal antibody specifically detects PTP1B/PTPN1 in Human samples. It is validated for Western Blot, ELISA, Immunoprecipitation

Specifications

Antigen PTP1B/PTPN1
Applications Western Blot, ELISA, Immunoprecipitation
Classification Monoclonal
Clone 226
Conjugate Janelia Fluor 549
Dilution Western Blot, ELISA, Immunoprecipitation
Formulation 50mM Sodium Borate
Gene Alias EC 3.1.3.48, protein tyrosine phosphatase 1B, protein tyrosine phosphatase, non-receptor type 1, Protein-tyrosine phosphatase 1B, PTP-1B, PTP1Bprotein tyrosine phosphatase, placental, tyrosine-protein phosphatase non-receptor type 1
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human PTP1B/PTPN1 (Accession#: NP_002818.1; Glu 2-Asn 321).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 5770
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.