Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PTP1B/PTPN1 Antibody, DyLight 488, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP128691G
Description
PTP1B/PTPN1 Polyclonal antibody specifically detects PTP1B/PTPN1 in Human, Rat samples. It is validated for Western BlotSpecifications
PTP1B/PTPN1 | |
Polyclonal | |
Western Blot | |
EC 3.1.3.48, protein tyrosine phosphatase 1B, protein tyrosine phosphatase, non-receptor type 1, Protein-tyrosine phosphatase 1B, PTP-1B, PTP1Bprotein tyrosine phosphatase, placental, tyrosine-protein phosphatase non-receptor type 1 | |
The immunogen recognized by this antibody maps to a region between residue 275 and 325 of human protein tyrosine phosphatase 1B using the numbering given in entry NP_002818.1 (GeneID 5770). | |
0.1 mL | |
Phospho Specific | |
5770 | |
Store at 4°C in the dark. | |
IgG |
Western Blot | |
DyLight 488 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction