Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PTP1B/PTPN1 Antibody (OTI1D10), PE/Atto594, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273729PEATT594
Description
PTP1B/PTPN1 Monoclonal specifically detects PTP1B/PTPN1 in Human, Rat, Canine, Monkey, Primate samples. It is validated for Flow Cytometry, Knockout Validated.Specifications
PTP1B/PTPN1 | |
Monoclonal | |
PE/ATTO 594 | |
PBS with 0.05% Sodium Azide | |
PTPN1 | |
Full length recombinant protein of human PTP1B/PTPN1 (NP_002818) produced in HEK293 cell. | |
0.1 mL | |
Primary | |
Human, Rat, Canine, Monkey, Primate | |
IgG2a |
Flow Cytometry, Immunoassay | |
OTI1D10 | |
Flow Cytometry, Knockout Validated | |
EC 3.1.3.48, protein tyrosine phosphatase 1B, protein tyrosine phosphatase, non-receptor type 1, Protein-tyrosine phosphatase 1B, PTP-1B, PTP1Bprotein tyrosine phosphatase, placental, tyrosine-protein phosphatase non-receptor type 1 | |
Mouse | |
Affinity Purified | |
Phospho Specific | |
5770.0 | |
Store at 4°C in the dark. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction