Learn More
Description
Specifications
Specifications
| Antigen | PTP1B/PTPN1 |
| Applications | Flow Cytometry, Immunoassay |
| Classification | Monoclonal |
| Clone | OTI1D10 |
| Conjugate | PE |
| Dilution | Flow Cytometry, Knockout Validated |
| Formulation | PBS with 0.05% Sodium Azide |
| Gene Alias | EC 3.1.3.48, protein tyrosine phosphatase 1B, protein tyrosine phosphatase, non-receptor type 1, Protein-tyrosine phosphatase 1B, PTP-1B, PTP1Bprotein tyrosine phosphatase, placental, tyrosine-protein phosphatase non-receptor type 1 |
| Gene Symbols | PTPN1 |
| Host Species | Mouse |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
