Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PTP1B/PTPN1, p Ser50 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP323195
Description
PTP1B/PTPN1 Polyclonal antibody specifically detects PTP1B/PTPN1 in Human, Mouse, Rat samples. It is validated for Western Blot, ImmunofluorescenceSpecifications
PTP1B/PTPN1 | |
Polyclonal | |
Western Blot 1:1000, Immunocytochemistry/Immunofluorescence 1:100 | |
EC 3.1.3.48, protein tyrosine phosphatase 1B, protein tyrosine phosphatase, non-receptor type 1, Protein-tyrosine phosphatase 1B, PTP-1B, PTP1Bprotein tyrosine phosphatase, placental, tyrosine-protein phosphatase non-receptor type 1 | |
PTP1B/PTPN1 (Ser-50) synthetic peptide (coupled to carrier protein) corresponds to amino acids surrounding Ser-50 in human PTP1B/PTPN1. This sequence has significant homology to the conserved site in rat and mouse PTP1B/PTPN1, and has homology to conserved sites in TC-PTP and MPTP, but not other protein tyrosine phosphatase family members. [UniProt# P18031] | |
0.1 mL | |
Phospho Specific | |
5770 | |
Store at -20°C. | |
IgG |
Western Blot, Immunofluorescence | |
Unconjugated | |
PBS, 1 mg/ml BSA and 50% glycerol | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction