Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PTP4A2 Monoclonal Antibody (5B6F4)
GREENER_CHOICE

Catalog No. PIMA531870
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA531870 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA531870 Supplier Invitrogen™ Supplier No. MA531870
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA5-31870 has been tested in indirect ELISA.

PRL-2, also known as Protein tyrosine phosphatase 4a2 (Ptp4a2) is a unique nuclear protein tyrosine phosphatase (PTP) that plays a central role in regulating diverse cellular processes. PRL-1 is induced in mitogen-stimulated cells and regenerating liver. PRL-2 exhibits 87% identity to PRL-1 in their amino acid sequences. All mouse PRL proteins contain a C-terminal consensus sequence for prenylation. All PRL proteins bear significant sequence homology to Cdc14p and the tumor suppressor PTEN/MMAC1. PRL-2 is preferentially expressed in skeletal muscle. PRL-2 is also expressed at lower levels in other tissues.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PTP4A2
Applications ELISA, Western Blot
Classification Monoclonal
Clone 5B6F4
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% sodium azide
Gene PTP4A2
Gene Accession No. Q12974
Gene Alias BM-008; HH13; HH7-2; HU-PP-1; OV-1; phosphatase of regenerating liver 2; PRL2; PRL-2; protein tyrosine phosphatase 4a2; protein tyrosine phosphatase IVA; protein tyrosine phosphatase IVA2; protein tyrosine phosphatase type IVA 2; protein tyrosine phosphatase type IVA, member 2; protein-tyrosine phosphatase 4a2; Protein-tyrosine phosphatase of regenerating liver 2; PTP(CAAXII); PTP4A; Ptp4a2; PTPCAAX2; ptp-IV1a; ptp-IV1b; tyrosine-phosphatase
Gene Symbols PTP4A2
Host Species Mouse
Immunogen Purified recombinant fragment of human PTP4A2 (AA: 58-162) expressed in E. coli.
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8073
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.