Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PTP4A2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5102987
Description
Antibody detects endogenous levels of total PTP4A2.
PRL-2, also known as Protein tyrosine phosphatase 4a2 (Ptp4a2) is a unique nuclear protein tyrosine phosphatase (PTP) that plays a central role in regulating diverse cellular processes. PRL-1 is induced in mitogen-stimulated cells and regenerating liver. PRL-2 exhibits 87% identity to PRL-1 in their amino acid sequences. All mouse PRL proteins contain a C-terminal consensus sequence for prenylation. All PRL proteins bear significant sequence homology to Cdc14p and the tumor suppressor PTEN/MMAC1. PRL-2 is preferentially expressed in skeletal muscle. PRL-2 is also expressed at lower levels in other tissues.
Specifications
PTP4A2 | |
Polyclonal | |
Unconjugated | |
PTP4A2 | |
BM-008; HH13; HH7-2; HU-PP-1; OV-1; phosphatase of regenerating liver 2; PRL2; PRL-2; protein tyrosine phosphatase 4a2; protein tyrosine phosphatase IVA; protein tyrosine phosphatase IVA2; protein tyrosine phosphatase type IVA 2; protein tyrosine phosphatase type IVA, member 2; protein-tyrosine phosphatase 4a2; Protein-tyrosine phosphatase of regenerating liver 2; PTP(CAAXII); PTP4A; Ptp4a2; PTPCAAX2; ptp-IV1a; ptp-IV1b; tyrosine-phosphatase | |
Rabbit | |
Affinity chromatography | |
RUO | |
8073 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q12974 | |
PTP4A2 | |
A synthesized peptide derived from human PTP4A2(Accession Q12974), corresponding to amino acid residues N28-V78. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction