Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PTPD1 Rabbit anti-Chicken, Human, Mouse, Non-human Primate, Rat, Polyclonal, FabGennix
Rabbit Polyclonal Antibody
Supplier: Fabgennix Inc PTPN101AP

Description
PTPD1 Polyclonal antibody specifically detects PTPD1 in Chicken, Human, Mouse, Non-human Primate, Rat samples. It is validated for ELISA, Immunohistochemistry, Immunoprecipitation, Western BlotSpecifications
PTPD1 | |
Polyclonal | |
Unconjugated | |
PTPN21 | |
dJ1175B15.2 (protein tyrosine phosphatase, non-receptor type 21); protein tyrosine phosphatase 2E; protein tyrosine phosphatase, non-receptor type 21; protein-tyrosine phosphatase 2E; Protein-tyrosine phosphatase D1; protein-tyrosine phosphatase PTP-RL10; Ptp2E; PTPD1; Ptpn21; PTPRL10; Tyrosine-protein phosphatase non-receptor type 21 | |
Synthetic peptide taken within amino acid region 400-500 on mouse PTPND1 protein. | |
200 μL | |
Primary | |
Chicken, Human, Mouse, Non-human Primate, Rat | |
Antibody | |
IgG |
ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot | |
0.5-1.5 mg/mL | |
proprietary buffer with 0.5% BSA, 30% glycerol and 0.02% sodium azide; pH 7.4-7.8 | |
Q16825, Q62136, Q62728 | |
Rabbit | |
Affinity chromatography | |
RUO | |
11099, 171070, 24000, 423398, 453090 | |
-20°C | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction