Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PTPDC1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA555433
Description
Immunogen sequence: GMIFSNEQQF DPLWKRRNVE CLQPLTHLKR RLSYSDSDLK RAENLLEQGE TPQTVPAQIL VGHKPRQQKL ISHCYIPQSP EPDLHKEALV RSTLSFWSQS KFGGLEGLKD NGSPIFHGRI IPKEAQQSGA FSADVSGSHS PGE Highest antigen sequence identity to the following orthologs: Mouse - 65%, Rat - 63%.

Specifications
PTPDC1 | |
Polyclonal | |
Unconjugated | |
PTPDC1 | |
protein tyrosine phosphatase domain containing 1; protein tyrosine phosphatase domain-containing protein 1; protein tyrosine phosphatase PTP9Q22; PTP9Q22; PTPDC1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
138639 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
A2A3K4 | |
PTPDC1 | |
Recombinant protein corresponding to Human PTPDC1. Recombinant protein control fragment (Product #RP-91563). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction