Learn More
PTPIP51 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
$151.31
Specifications
Antigen | PTPIP51 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Liquid |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-156-3584
![]() |
Bethyl Laboratories, Inc
A302829AT |
20 μL |
Each of 1 for $151.31
|
|
|||||
Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Orangutan
RMDN3 is involved in cellular calcium homeostasis regulation. It may participate in differentiation and apoptosis of keratinocytes. Its overexpression induces apoptosis.Specifications
PTPIP51 | |
Polyclonal | |
Liquid | |
RUO | |
TBS with 0.1% BSA and 0.09% sodium azide | |
cerebral protein 10, FAM82A2, FAM82C, family with sequence similarity 82, member A2, family with sequence similarity 82, member C, FLJ10579, hRMD-3, microtubule-associated protein, Protein FAM82A2, Protein FAM82C, protein tyrosine phosphatase-interacting protein 51, PTPIP51, RMD3, RMD-3, TCPTP-interacting protein 51 | |
RMDN3 | |
IgG | |
Antigen affinity chromatography | |
Antibody |
Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
Q96TC7 | |
55177 | |
Between 420 and 470 | |
Primary | |
4° C | |
RMDN3 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.