Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PTPIP51 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501986747
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-6747 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-6747 Supplier Bioss Supplier No. BS5719R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

RMDN3 is a protein coding gene involved in cellular calcium homeostasis regulation. May participate in differentiation and apoptosis of keratinocytes. Overexpression induces apoptosis.

Specifications

Antigen PTPIP51
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene RMDN3
Gene Accession No. Q96TC7
Gene Alias 1200015F23Rik; AI131757; BOS_10730; Cerebral protein 10; Fam82a2; Fam82c; family with sequence similarity 82, member A2; family with sequence similarity 82, member C; FLJ10579; hRMD-3; hucep-10; LOC100345867; microtubule-associated protein; Protein FAM82A2; Protein FAM82C; Protein tyrosine phosphatase-interacting protein 51; PTPIP51; regulator of microtubule dynamics 3; Regulator of microtubule dynamics protein 3; RGD1308697; Rmd3; RMD-3; RMDN3; TCPTP-interacting protein 51; tyrosine phosphatase interacting protein 51; UNQ3122/PRO10274
Gene Symbols RMDN3
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human PTPIP51/FAM82A2.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55177
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.