Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PTPLAD1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP159028
Description
PTPLAD1 Polyclonal specifically detects PTPLAD1 in Human samples. It is validated for Western Blot.Specifications
PTPLAD1 | |
Polyclonal | |
Unconjugated | |
PBS, 2% Sucrose with 0.09% Sodium Azide | |
PTPLAD1 | |
Synthetic peptides corresponding to PTPLAD1(protein tyrosine phosphatase-like A domain containing 1) The peptide sequence was selected from the N terminal of PTPLAD1. Peptide sequence WLDESDAEMELRAKEEERLNKLRLESEGSPETLTNLRKGYLFMYNLVQFL. | |
100 μL | |
Signal Transduction | |
51495 | |
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Western Blot | |
0.5 mg/ml | |
Western Blot 1.0 ug/ml | |
B-ind1, BIND1, Butyrate-induced protein 1, butyrate-induced transcript 1, EC 4.2.1.-, FLJ90376, HACD3, hB-ind1, HSPC121,3-hydroxyacyl-CoA dehydratase 3, protein tyrosine phosphatase-like A domain containing 1, Protein tyrosine phosphatase-like protein PTPLAD1, Protein-tyrosine phosphatase-like A domain-containing protein 1 | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Expected identity based on immunogen sequence: Guinea pig: 100%; Equine: 100%; Human: 100%; Pig: 100%; Rabbit: 100%; Canine: 92%. | |
Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction