Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PTPLB Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5113050
Description
PTPLB Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA
PTPLB is probable anti-phosphatase.
Specifications
PTPLB | |
Polyclonal | |
Unconjugated | |
Hacd2 | |
3-hydroxyacyl-CoA dehydratase 2; 6330408J20Rik; AI255777; AI481689; HACD2; Hcad2; I79_000662; protein tyrosine phosphatase-like (proline instead of catalytic arginine), member b; protein-tyrosine phosphatase-like member B; Ptplb; very-long-chain (3R)-3-hydroxyacyl-[acyl-carrier protein] dehydratase 2; very-long-chain (3R)-3-hydroxyacyl-CoA dehydratase 2 | |
Rabbit | |
Protein G | |
RUO | |
201562 | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
Q6Y1H2 | |
Hacd2 | |
Recombinant Human Very-long-chain (3R)-3-hydroxyacyl-CoA dehydratase 2 protein (7-41AA). | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction