Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PTPMT1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560557
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560557 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560557 Supplier Invitrogen™ Supplier No. PA560557
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: IPTLDNLQKG VQFALKYQSL GQCVYVHCKA GRSRSATMVA AYLIQVHKWS PEEAVRAIAK IRSYIHIRPG QLDVLKEFHK QITARAT Highest antigen sequence identity to the following orthologs: Mouse - 85%, Rat - 83%.

Lipid phosphatase which dephosphorylates phosphatidylglycerophosphate (PGP) to phosphatidylglycerol (PG). PGP is an essential intermediate in the biosynthetic pathway of cardiolipin, a mitochondrial-specific phospholipid regulating the membrane integrity and activities of the organelle. Has also been shown to display phosphatase activity toward phosphoprotein substrates, specifically mediates dephosphorylation of mitochondrial proteins, thereby playing an essential role in ATP production. Has probably a preference for proteins phosphorylated on Ser and/or Thr residues compared to proteins phosphorylated on Tyr residues. Probably involved in regulation of insulin secretion in pancreatic beta cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PTPMT1
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PTPMT1
Gene Accession No. Q8WUK0
Gene Alias 1110001D10Rik; 2810004N20Rik; DUSP23; MOSP; NB4 apoptosis/differentiation related protein; phosphatidylglycerophosphatase and protein-tyrosine phosphatase 1; Phosphoinositide lipid phosphatase; Plip; PNAS-129; protein tyrosine phosphatase, mitochondrial 1; protein-tyrosine phosphatase mitochondrial 1; PTEN-like phosphatase; PTPMT1
Gene Symbols PTPMT1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PTPMT1. Recombinant protein control fragment (Product #RP-97310).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 114971
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.