Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PTPMT1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA550779
Description
The antibody detects endogenous levels of total PTPMT1 protein.
Lipid phosphatase which dephosphorylates phosphatidylglycerophosphate (PGP) to phosphatidylglycerol (PG). PGP is an essential intermediate in the biosynthetic pathway of cardiolipin, a mitochondrial-specific phospholipid regulating the membrane integrity and activities of the organelle. Has also been shown to display phosphatase activity toward phosphoprotein substrates, specifically mediates dephosphorylation of mitochondrial proteins, thereby playing an essential role in ATP production. Has probably a preference for proteins phosphorylated on Ser and/or Thr residues compared to proteins phosphorylated on Tyr residues. Probably involved in regulation of insulin secretion in pancreatic beta cells.
Specifications
PTPMT1 | |
Polyclonal | |
Unconjugated | |
PTPMT1 | |
1110001D10Rik; 2810004N20Rik; DUSP23; MOSP; NB4 apoptosis/differentiation related protein; phosphatidylglycerophosphatase and protein-tyrosine phosphatase 1; Phosphoinositide lipid phosphatase; Plip; PNAS-129; protein tyrosine phosphatase, mitochondrial 1; protein-tyrosine phosphatase mitochondrial 1; PTEN-like phosphatase; PTPMT1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
114971 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
Q8WUK0 | |
PTPMT1 | |
Fusion protein corresponding to a region derived from internal residues of human protein tyrosine phosphatase, mitochondrial 1. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction