Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PTPN1 Mouse anti-Canine, Human, Clone: OTI1D10, lyophilized, TrueMAB™
SDP

Catalog No. 501662042
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-166-2042 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-166-2042 Supplier OriGene Supplier No. CF503310
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO

Ferritin is a ubiquitous and highly conserved protein which plays a major role in iron homeostasis. It is a holoenzyme shell (∽450 kDa) consisting of 24 subunits of two types, H (heavy) and L (light), and capable of storing up to 4,500 atoms of ferric iron. Depending on the tissue type and physiologic status of the cell, the ratio of H to L subunits in ferritin can vary widely. It can be viewed not only as part of a group of iron regulatory proteins that include transferrin and the transferrin receptor, but also as a member of the protein family that orchestrates the cellular defense against stress and inflammation. Ferritin is found in the liver, spleen, kidney and heart. Only a small amount is found in the blood. The blood level of ferritin serves as an indicator of the amount of iron stored in the body.

Specifications

Antigen PTPN1
Applications Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI1D10
Conjugate Unconjugated
Formulation PBS with 8% trehalose and no preservative; pH 7.3
Gene PTPN1
Gene Accession No. P18031
Gene Alias PTP1B
Gene Symbols PTPN1
Host Species Mouse
Immunogen Full length human recombit protein of human PTPN1 produced in HEK293 cell.
Purification Method Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 477263, 5770
Target Species Canine, Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.