Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PTPN12 Antibody, DyLight 350, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP299820UV
Description
PTPN12 Polyclonal specifically detects PTPN12 in Human samples. It is validated for ELISA.Specifications
PTPN12 | |
Polyclonal | |
50 mM Sodium Borate | |
Rabbit | |
Affinity purified | |
RUO | |
5782 | |
Store at 4°C in the dark. | |
IgG |
ELISA | |
DyLight 350 | |
protein tyrosine phosphatase, non-receptor type 12, Protein-tyrosine phosphatase G1, PTPG1tyrosine-protein phosphatase non-receptor type 12, PTP-PESTEC 3.1.3.48 | |
Produced in rabbits immunized with purified, recombinant PTPN12 (AAA36529.1; Met1-Gln355). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction